siRNA Details
Gene Symbol: Gabra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017295.1 | NP_058991.1 | 7, 8 | 875 | ||
GenBank mRNA | X51992.1 | - | 7, 8 | 902 |

More Information
Other Species Targeted

Gene Ontology Categories:
