siRNA Details
Gene Symbol: Gabrg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183327.1 | NP_899156.1 | 4, 5 | 547 | ||
XM_006246125.2 | XP_006246187.1 | 4, 5 | 1110 | |||
GenBank mRNA | AY574252.1 | - | 4, 5 | 547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
