siRNA Details
Gene Symbol: Kcnj10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031602.1 | NP_113790.1 | 2 | 1159 | ||
XM_006250285.2 | XP_006250347.1 | 2 | 2445 | |||
XM_008769737.1 | XP_008767959.1 | 2 | 1885 | |||
GenBank mRNA | U27558.1 | - | 2 | 1159 | ||
X83585.1 | - | 1 | 1036 | |||
X86818.1 | - | 2 | 1154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
