siRNA Details
Gene Symbol: Sema4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019272.1 | NP_062145.1 | 4 | 523 | ||
GenBank mRNA | AB002563.1 | - | 4 | 523 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011350, NM_001113481, AK047067, AF038652, AB021291, and others.
Gene Ontology Categories:
