siRNA Details
Gene Symbol: Atp5g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017311.1 | NP_059007.1 | 2, 3 | 95 | ||
XM_006247188.2 | XP_006247250.1 | 2, 3 | 138 | |||
XM_006247189.1 | XP_006247251.1 | 2, 3 | 243 | |||
GenBank mRNA | D13123.1 | - | 2, 3 | 95 | ||
FQ209790.1 | - | 2, 3 | 113 | |||
FQ210883.1 | - | 1, 2 | 59 | |||
FQ216886.1 | - | 2, 3 | 113 | |||
FQ217997.1 | - | 2, 3 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
