siRNA Details
Gene Symbol: Ubc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017314.1 | NP_059010.1 | 2425 | |||
GenBank mRNA | BC089218.1 | - | 1 | 1931 | ||
BC103477.1 | - | 3 | 1310 | 7929586 | ||
D17296.1 | - | 2425 | ||||
FQ212515.1 | - | 1 | 948 | |||
FQ229492.1 | - | 1 | 893 | |||
X92660.1 | - | 1 | 16 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
