siRNA Details
Gene Symbol: Hspb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031607.1 | NP_113795.1 | 3 | 790 | ||
GenBank mRNA | BC089930.1 | - | 1 | 242 | ||
BC169050.1 | - | 3 | 778 | 7103781 | ||
CK358485.1 | - | 2 | 528 |
