siRNA Details
Gene Symbol: Ptk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017318.2 | NP_059014.2 | 11 | 1150 | ||
XM_006252143.2 | XP_006252205.1 | 11 | 1475 | |||
XM_006252144.2 | XP_006252206.1 | 11 | 4466 | |||
XM_006252145.2 | XP_006252207.1 | 11 | 1279 | |||
XM_008770773.1 | XP_008768995.1 | 11 | 2663 | |||
XM_008770774.1 | XP_008768996.1 | 11 | 2229 | |||
GenBank mRNA | BC101921.1 | - | 11 | 1150 | 7937929 | |
D45854.1 | - | 11 | 1328 | |||
U69109.1 | - | 10 | 1070 |
