siRNA Details
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017335.1 | NP_059031.1 | 14 | 1915 | ||
GenBank mRNA | FQ229573.1 | - | 1439 | |||
U28927.1 | - | 14 | 1915 |

More Information
Other Species Targeted

Gene Ontology Categories:
