siRNA Details
Gene Symbol: Hpcal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017356.2 | NP_059052.1 | 4 | 777 | ||
XM_006239914.2 | XP_006239976.1 | 5 | 1384 | |||
XM_006239916.2 | XP_006239978.1 | 4 | 1279 | |||
XM_008764605.1 | XP_008762827.1 | 4 | 894 | |||
XM_008764606.1 | XP_008762828.1 | 6 | 1494 | |||
GenBank mRNA | BC088759.1 | - | 4 | 777 | 7317905 | |
D13126.1 | - | 4 | 732 | |||
FQ214068.1 | - | 2 | 300 | |||
FQ220970.1 | - | 4 | 767 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
