siRNA Details
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017359.2 | NP_059055.2 | 1, 2 | 184 | ||
GenBank mRNA | BC085744.1 | - | 1, 2 | 184 | 7191921 |

More Information
Other Species Targeted

Gene Ontology Categories:
