siRNA Details
Gene Symbol: Chrm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017362.4 | NP_059058.1 | 1357 | |||
GenBank mRNA | M22926.1 | - | 1357 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
