siRNA Details
Gene Symbol: LOC103689989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771673.1 | XP_008769895.1 | 619 | |||
XM_008771674.1 | XP_008769896.1 | 657 |
Gene Symbol: LOC100911206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257188.2 | XP_006257250.2 | 3 | 400 |
Gene Symbol: LOC103690150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771672.1 | XP_008769894.1 | 657 | |||
GenBank mRNA | FQ228618.1 | - | 680 |
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253914.2 | 5 | 680 | |||
GenBank mRNA | FQ220053.1 | - | 684 | |||
FQ220072.1 | - | 680 | ||||
FQ220274.1 | - | 682 | ||||
FQ220568.1 | - | 682 | ||||
FQ220587.1 | - | 680 | ||||
FQ220864.1 | - | 406 | ||||
FQ221231.1 | - | 682 | ||||
FQ222593.1 | - | 680 | ||||
FQ228787.1 | - | 680 | ||||
FQ229328.1 | - | 680 | ||||
FQ229537.1 | - | 679 | ||||
FQ229656.1 | - | 674 | ||||
FQ230119.1 | - | 680 | ||||
M55269.1 | - | 673 |
Gene Symbol: Prl3d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771671.1 | XP_008769893.1 | 5 | 683 | ||
GenBank mRNA | BC161851.1 | - | 5 | 657 | 9088452 | |
D21103.1 | - | 5 | 619 | |||
DQ329282.1 | - | 5 | 676 | |||
DQ329283.1 | - | 5 | 676 | |||
FQ145043.1 | - | 5 | 682 | |||
FQ221365.1 | - | 5 | 680 | |||
FQ223255.1 | - | 5 | 683 | |||
FQ223518.1 | - | 5 | 680 | |||
FQ229928.1 | - | 5 | 682 | |||
FQ230035.1 | - | 5 | 680 |
Gene Symbol: Prl3d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033233.1 | NP_150236.1 | 5 | 637 | ||
XM_008771595.1 | XP_008769817.1 | 5 | 619 | |||
GenBank mRNA | BC097302.1 | - | 5 | 663 | 7444366 | |
U32679.1 | - | 5 | 637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
