siRNA Details
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017365.2 | NP_059061.1 | 2 | 184 | ||
GenBank mRNA | BC072465.1 | - | 2 | 175 | 7100446 | |
U23769.1 | - | 2 | 184 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdlim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017365.2 | NP_059061.1 | 2 | 184 | ||
GenBank mRNA | BC072465.1 | - | 2 | 175 | 7100446 | |
U23769.1 | - | 2 | 184 |