siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019124.1 | NP_061997.1 | 9 | 1310 | ||
XM_006246795.2 | XP_006246857.1 | 9 | 1473 | |||
XM_006246796.2 | XP_006246858.1 | 9 | 1473 | |||
XM_008767861.1 | XP_008766083.1 | 9 | 2104 | |||
GenBank mRNA | D85844.1 | - | 9 | 1310 | ||
U70777.1 | - | 9 | 1457 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019400, AK137236, D86066, AB001750, AF248489, and others.
Gene Ontology Categories:
