siRNA Details
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019124.1 | NP_061997.1 | 9 | 1445 | ||
XM_006246795.2 | XP_006246857.1 | 9 | 1608 | |||
XM_006246796.2 | XP_006246858.1 | 9 | 1608 | |||
XM_008767861.1 | XP_008766083.1 | 9 | 2239 | |||
GenBank mRNA | D85844.1 | - | 9 | 1445 | ||
U70777.1 | - | 9 | 1592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
