siRNA Details
Gene Symbol: Cacna1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019294.2 | NP_062167.2 | 27, 28 | 3942 | ||
XM_006250069.2 | XP_006250131.1 | 30, 31 | 5316 | |||
GenBank mRNA | AY029412.1 | - | 27, 28 | 3942 | ||
L15453.1 | - | 28, 29 | 4039 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
