siRNA Details
Gene Symbol: Cr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005265.1 | NP_001005265.1 | 5 | 896 | ||
NM_001005330.1 | NP_001005330.1 | 896 | ||||
NM_019301.2 | NP_062174.1 | 5 | 896 | |||
GenBank mRNA | BC061736.1 | - | 5 | 665 | 5599318 | |
D42114.1 | - | 5 | 705 | |||
D42115.1 | - | 5 | 641 | |||
D42116.1 | - | 2 | 249 | |||
L36532.1 | - | 5 | 664 |
