siRNA Details
Gene Symbol: Cr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005265.1 | NP_001005265.1 | 4, 5 | 720 | ||
NM_001005330.1 | NP_001005330.1 | 720 | ||||
NM_019301.2 | NP_062174.1 | 4, 5 | 720 | |||
GenBank mRNA | BC061736.1 | - | 4, 5 | 489 | 5599318 | |
D42114.1 | - | 4, 5 | 529 | |||
D42115.1 | - | 4, 5 | 465 | |||
D42116.1 | - | 1, 2 | 73 | |||
L19118.1 | - | 1, 2 | 83 | |||
L36532.1 | - | 4, 5 | 488 |
