siRNA Details
Gene Symbol: Crk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019302.1 | NP_062175.1 | 3 | 850 | ||
XM_006246913.1 | XP_006246975.1 | 3 | 858 | |||
GenBank mRNA | D44481.1 | - | 3 | 850 | ||
FQ210506.1 | - | 3 | 863 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001277219, NM_001277221, NM_133656, BC012216, AK199005, and others.
Gene Ontology Categories:
