siRNA Details
Gene Symbol: Cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077642.1 | NP_001071110.1 | 4 | 513 | ||
GenBank mRNA | M92059.1 | - | 4 | 510 | ||
S73894.1 | - | 4 | 513 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
