siRNA Details
Gene Symbol: Fgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019305.2 | NP_062178.1 | 3 | 918 | ||
GenBank mRNA | M22427.1 | - | 918 | |||
X07285.1 | - | 3 | 639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
