siRNA Details
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022927.1 | NP_075216.1 | 3 | 972 | ||
XM_006256825.2 | XP_006256887.1 | 3 | 2256 | |||
GenBank mRNA | AF186461.1 | - | 3 | 972 |

More Information
Other Species Targeted

Gene Ontology Categories:
