siRNA Details
Gene Symbol: Gata4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144730.1 | NP_653331.1 | 5 | 1524 | ||
XM_006252189.2 | XP_006252251.1 | 4 | 1104 | |||
GenBank mRNA | L22761.1 | - | 5 | 1524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
