siRNA Details
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105731.3 | NP_001099201.3 | 17 | 1945 | ||
GenBank mRNA | EU251189.1 | - | 17 | 1945 |

More Information
Other Species Targeted

Gene Ontology Categories:
