siRNA Details
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105731.3 | NP_001099201.3 | 10, 11 | 1276 | ||
GenBank mRNA | EU251189.1 | - | 10, 11 | 1276 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Prkg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105731.3 | NP_001099201.3 | 10, 11 | 1276 | ||
GenBank mRNA | EU251189.1 | - | 10, 11 | 1276 |