siRNA Details
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019335.1 | NP_062208.1 | 10 | 925 | ||
XM_008764425.1 | XP_008762647.1 | 9 | 1149 | |||
XM_008764426.1 | XP_008762648.1 | 10 | 991 | |||
GenBank mRNA | L29281.1 | - | 10 | 925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
