siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 11 | 2350 | ||
XM_006238260.2 | XP_006238322.1 | 18 | 2933 | |||
XM_008763800.1 | XP_008762022.1 | 7 | 1736 | |||
XM_008763801.1 | XP_008762023.1 | 2 | 1253 | |||
XM_008763802.1 | XP_008762024.1 | 2 | 2692 | |||
GenBank mRNA | AB055153.1 | - | 11 | 2350 |

More Information
Other Species Targeted

Gene Ontology Categories:
