siRNA Details
Gene Symbol: Rgs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019340.1 | NP_062213.1 | 3 | 235 | ||
XM_006238260.2 | XP_006238322.1 | 10 | 818 | |||
XM_006238263.2 | XP_006238325.1 | 12 | 993 | |||
XM_006238264.2 | XP_006238326.1 | 10 | 820 | |||
XM_006238265.2 | XP_006238327.1 | 3 | 2435 | |||
GenBank mRNA | AB055153.1 | - | 3 | 235 | ||
BC085710.1 | - | 3 | 224 |
