siRNA Details
Gene Symbol: SNTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006750.3 | NP_006741.1 | 2 | 626 | ||
GenBank mRNA | AF243385.1 | - | 2 | 626 | ||
BC048215.1 | - | 2 | 611 | 5805707 | ||
U40572.1 | - | 2 | 626 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009229, AK165009, BC145442, AK080059, AK040423, and others.
Gene Ontology Categories:
