siRNA Details
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019353.2 | NP_062226.2 | 16 | 2653 | ||
XM_008764607.1 | XP_008762829.1 | 15 | 2611 | |||
XM_008764608.1 | XP_008762830.1 | 16 | 2647 | |||
GenBank mRNA | M31655.1 | - | 12 | 2179 | ||
X17396.1 | - | 16 | 2652 |
