siRNA Details
Gene Symbol: Tpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019353.2 | NP_062226.2 | 11 | 1962 | ||
XM_008764607.1 | XP_008762829.1 | 10 | 1920 | |||
XM_008764608.1 | XP_008762830.1 | 11 | 1956 | |||
GenBank mRNA | M31655.1 | - | 7 | 1488 | ||
X17396.1 | - | 11 | 1961 |
