siRNA Details
Gene Symbol: Eif2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019356.1 | NP_062229.1 | 3 | 318 | ||
GenBank mRNA | BC087019.1 | - | 3 | 375 | 7191037 | |
J02646.1 | - | 3 | 318 |

More Information
Other Species Targeted

Gene Ontology Categories:
