siRNA Details
Gene Symbol: Cox6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019360.2 | NP_062233.2 | 4 | 385 | ||
GenBank mRNA | BC058480.1 | - | 4 | 361 | 6922382 | |
CF976717.1 | - | 4 | 389 | |||
FQ210091.1 | - | 4 | 363 | |||
FQ210263.1 | - | 4 | 362 | |||
FQ217805.1 | - | 4 | 388 | |||
FQ222319.1 | - | 4 | 357 | |||
FQ222901.1 | - | 4 | 406 | |||
FQ223820.1 | - | 4 | 360 | |||
K01565.1 | - | 2 | 172 | |||
M27466.1 | - | 4 | 396 | |||
M27467.1 | - | 3 | 360 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
