siRNA Details
Gene Symbol: Arc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019361.1 | NP_062234.1 | 1 | 295 | ||
XM_008765590.1 | XP_008763812.1 | 1 | 808 | |||
XM_008765591.1 | XP_008763813.1 | 1 | 808 | |||
GenBank mRNA | U19866.1 | - | 1 | 295 | ||
Z46925.1 | - | 1 | 276 |

More Information
Other Species Targeted

Gene Ontology Categories:
