siRNA Details
Gene Symbol: Scfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019364.1 | NP_062237.1 | 11 | 951 | ||
XM_006240085.2 | XP_006240147.1 | 11 | 1000 | |||
GenBank mRNA | D79221.1 | - | 11 | 951 | ||
U35364.1 | - | 11 | 971 | |||
U57687.1 | - | 11 | 938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
