siRNA Details
Gene Symbol: Scfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019364.1 | NP_062237.1 | 17 | 1469 | ||
XM_006240085.2 | XP_006240147.1 | 17 | 1518 | |||
GenBank mRNA | D79221.1 | - | 17 | 1469 | ||
U35364.1 | - | 17 | 1489 | |||
U57687.1 | - | 17 | 1456 |

More Information
Other Species Targeted

Gene Ontology Categories:
