siRNA Details
Gene Symbol: Bet1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019368.3 | NP_062241.2 | 3 | 179 | ||
XM_006230574.2 | XP_006230636.1 | 4 | 391 | |||
GenBank mRNA | AF003998.1 | - | 3 | 218 | ||
BC059138.1 | - | 3 | 183 | 6890305 | ||
FQ228853.1 | - | 4 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
