siRNA Details
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019369.2 | NP_062242.2 | 6, 7 | 772 | ||
GenBank mRNA | BC089806.1 | - | 6, 7 | 772 | 7326076 | |
FQ210475.1 | - | 6, 7 | 728 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019369.2 | NP_062242.2 | 6, 7 | 772 | ||
GenBank mRNA | BC089806.1 | - | 6, 7 | 772 | 7326076 | |
FQ210475.1 | - | 6, 7 | 728 |