siRNA Details
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019369.2 | NP_062242.2 | 4 | 445 | ||
GenBank mRNA | BC089806.1 | - | 4 | 445 | 7326076 | |
FQ210475.1 | - | 4 | 401 | |||
Y11283.1 | - | 4 | 442 |

More Information
Other Species Targeted

Gene Ontology Categories:
