siRNA Details
Gene Symbol: Enpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019370.2 | NP_062243.2 | 17 | 1606 | ||
GenBank mRNA | BC097326.1 | - | 17 | 1606 | 7458431 | |
D30649.1 | - | 17 | 1633 | |||
U78787.1 | - | 17 | 1603 | |||
U78788.1 | - | 17 | 1609 | |||
Z47987.1 | - | 17 | 1590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
