siRNA Details
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019385.2 | NP_062258.2 | 5 | 473 | ||
GenBank mRNA | AF015264.2 | - | 5 | 450 | ||
BC081809.1 | - | 5 | 467 | |||
BC161850.1 | - | 5 | 473 | 8369057 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
