siRNA Details
Gene Symbol: Cyp4f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019623.2 | NP_062569.2 | 10 | 1391 | ||
XM_008765180.1 | XP_008763402.1 | 11 | 1342 | |||
XM_008765181.1 | XP_008763403.1 | 11 | 1285 | |||
GenBank mRNA | BC081808.1 | - | 10 | 1391 | 7109036 | |
FQ218792.1 | - | 5 | 571 | |||
M94548.1 | - | 10 | 1284 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
