siRNA Details
Gene Symbol: Mrpl37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004235.1 | NP_001004235.1 | 4 | 819 | ||
GenBank mRNA | BC079328.1 | - | 4 | 819 | 7191681 | |
FQ215540.1 | - | 4 | 832 | |||
FQ226708.1 | - | 2 | 199 | |||
FQ231717.1 | - | 2 | 199 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
