siRNA Details
Gene Symbol: Plvap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020086.1 | NP_064471.1 | 3 | 789 | ||
GenBank mRNA | AF154831.1 | - | 3 | 789 | ||
BC070900.1 | - | 3 | 789 | 7100403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
