siRNA Details
Gene Symbol: Ndufaf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033971.1 | NP_001029143.1 | 2 | 884 | ||
NM_020080.2 | NP_064465.1 | 2 | 902 | |||
XM_006243845.2 | XP_006243907.1 | 3 | 2384 | |||
GenBank mRNA | BC079306.1 | - | 2 | 636 | 7134644 | |
FQ216095.1 | - | 2 | 253 | |||
FQ223486.1 | - | 2 | 197 | |||
FQ233128.1 | - | 1 | 329 | |||
U95160.1 | - | 2 | 238 | |||
U95161.1 | - | 2 | 238 | |||
U95162.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
