siRNA Details
Gene Symbol: Acpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134901.1 | NP_001128373.1 | 2 | 151 | ||
NM_020072.1 | NP_064457.1 | 2 | 191 | |||
GenBank mRNA | DQ826426.1 | - | 2 | 151 | ||
M32397.1 | - | 2 | 191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
