siRNA Details
Gene Symbol: Adam17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020306.2 | NP_064702.1 | 9 | 1267 | ||
GenBank mRNA | AJ012603.1 | - | 9 | 1269 |

More Information
Other Species Targeted

Gene Ontology Categories:
