siRNA Details
Gene Symbol: Cnr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020543.4 | NP_065418.3 | 3 | 1155 | ||
GenBank mRNA | AF218846.1 | - | 3 | 1155 | ||
BC107941.1 | - | 1 | 1290 | |||
FJ694960.1 | - | 3 | 2409 | |||
FJ694961.1 | - | 3 | 2511 | |||
JN420349.1 | - | 4 | 2606 | |||
JX494784.1 | - | 4 | 1725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
