siRNA Details
Gene Symbol: Nt5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021576.2 | NP_067587.2 | 2, 3 | 612 | ||
GenBank mRNA | BC081806.1 | - | 2, 3 | 642 | 7134340 | |
BC098665.1 | - | 2, 3 | 612 | 7322840 | ||
J05214.1 | - | 2, 3 | 962 |
